Skip to Content
Merck
All Photos(1)

Key Documents

EHU099881

Sigma-Aldrich

MISSION® esiRNA

targeting human FN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACACCTTCGGGGGAAATAATTCCTGTGAATATTCTTTTTCAATTCAGCAAACATTTGAAAATCTATGATGTGCAAGTCTAATTGTTGATTTCAGTACAAGATTTTCTAAATCAGTTGCTACAAAAACTGATTGGTTTTTGTCACTTCATCTCTTCACTAATGGAGATAGCTTTACACTTTCTGCTTTAATAGATTTAAGTGGACCCCAATATTTATTAAAATTGCTAGTTTACCGTTCAGAAGTATAATAGAAATAATCTTTAGTTGCTCTTTTCTAACCATTGTAATTCTTCCCTTCTTCCCTCCACCTTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuye Yu et al.
Oncogene, 39(27), 5042-5055 (2020-06-11)
Guanylate-binding protein 2 (GBP2) is an interferon-inducible large GTPase which is crucial to the protective immunity against microorganisms. However, its biological function in cancer remains largely unknown. Glioblastoma multiforme (GBM) is the most common and deadly brain tumor in adults.
Wenzhong Yi et al.
Oncology reports, 36(6), 3145-3153 (2016-10-18)
Fibronectin is a glycoprotein of the extracellular matrix, and regulates the processes of self-renewal and cell cycle progression. This study aimed to investigate fibronectin expression in colorectal cancer (CRC) and elucidate the effects of fibronectin on CRC by using a
Najla El-Hachem et al.
Cell death and differentiation, 25(11), 2010-2022 (2018-03-09)
HACE1 is an E3 ubiquitin ligase described as a tumour suppressor because HACE1-knockout mice develop multi-organ, late-onset cancers and because HACE1 expression is lost in several neoplasms, such as Wilms' tumours and colorectal cancer. However, a search of public databases
Changgeng Xu et al.
Molecular medicine reports, 13(1), 901-908 (2015-12-10)
Lefty is a member of the transforming growth factor (TGF) β superfamily, which is implicated in left‑right patterning during embryogenesis. Previous studies revealed that lefty attenuates the epithelial‑mesenchymal transition in tubular epithelial cells. In the present study, the protective effect
Ulrich Blache et al.
EMBO reports, 19(8) (2018-07-04)
The fate of mesenchymal stem cells (MSCs) in the perivascular niche, as well as factors controlling their fate, is poorly understood. Here, we study MSCs in the perivascular microenvironment of endothelial capillaries by modifying a synthetic 3D biomimetic poly(ethylene glycol)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service