CLS432065
Corning® CoolRack®
VS13, capacity 9 x 13 x 75 mm blood tubes
Synonym(s):
Corning Coolcell accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
foamed PE
feature
wells 9
Holds 9 x blood tubes 13x75 mm
packaging
pkg of 1
manufacturer/tradename
Corning 432065
technique(s)
cryopreservation: suitable
L × W × H
8.9 cm × 8.9 cm × 6.1 cm
size
26.7 mm , column spacing
26.7 mm , row spacing
well depth
61 mm
well diam.
13 mm
Looking for similar products? Visit Product Comparison Guide
General description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service