Skip to Content
Merck
All Photos(1)

Key Documents

MFE-280

98050131, human endometrium, Epithelial

Synonym(s):

MFE280 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MFE-280, 98050131

biological source

human endometrium

growth mode

Adherent

karyotype

Near diploid

morphology

Epithelial

products

Placental protein 14

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human caucasian endometrial adenocarcinoma

Cell Line Description

MFE-280 was derived from a recurrent, poorly differentiated endometrial carcinoma from a 78 year old patient. Initially, cells were grown as aggregates in suspension for one year, after which they were transferred to monolayer culture. The karyotype analysis revealed near-diploidy with a complex heterogeneous aberration pattern. MFE-280 were found to be positive for cytokines 7, 8, 18 and 19 as well as for vimentin. In addition, the cells express the placental protein 14, PP14. They have been shown to be tumorigenic in nude mice.

Application

Tumour biology studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,12
D13S317: 10,12
D16S539: 12
D5S818: 11,12
D7S820: 10
THO1: 7
TPOX: 9,10
vWA: 17

Culture Medium

MEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding 2-4 x 10,000 cells / cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service