Skip to Content
Merck
All Photos(1)

Key Documents

COV362

7071910, human ovary, Polygonal & fusiform

Synonym(s):

COV-362 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

COV362, 07071910

biological source

human ovary

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal & fusiform

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian epithelial-endometroid carcinoma

Cell Line Description

A human ovarian epithelial-endometroid carcinoma cell line established from a pleural effusion. Anchorage-independent growth in agar has been reported. The cell line COV362.4 (Sigma cat no. 07071904) is a subclone of this cell line.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 11,12
D13S317: 14
D16S539: 11
D5S818: 13
D7S820: 10,14
THO1: 9.3
TPOX: 11
vWA: 15,18

Culture Medium

DMEM + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service