Key Documents
AXYMTS06C
MiniTube
individual, bulk pack, size 0.65 mL, pkg of 5x960tips/cs, polypropylene, non-sterile
Synonym(s):
centrifuge tubes
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polypropylene
sterility
non-sterile
manufacturer/tradename
Axygen Inc MTS-06-C
Corning
packaging
pkg of 5x960tips/cs
size
0.65 mL
Features and Benefits
  • Ensures positive sample identification due to one-way lid design, in conjunction with α-numeric grid
  • Compatible with Tecan®, Beckman Coulter, ANSI/SLAS 1-2004, microplates, and other robotic workstations
  • DNase/RNase free and non-pyrogenic in nature
Legal Information
Corning is a registered trademark of Corning, Inc.
Coulter is a trademark of Beckman Coulter, Inc.
Tecan is a registered trademark of Tecan Group AG