Skip to Content
Merck
All Photos(3)

Key Documents

AV42117

Sigma-Aldrich

Anti-TNNI1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp451O223, Anti-TNN1, Anti-Troponin I type 1 (skeletal, slow)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

rat, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNNI1(7135)

Immunogen

Synthetic peptide directed towards the N terminal region of human TNNI1

Biochem/physiol Actions

Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Each troponin-tropomyosin complex contains 1 molecule from each of the 3 troponins: T, C, and I. The troponin I (TNI) subfamily contains three genes: TNI-skeletal-fast-twitch, TNI-skeletal-slow-twitch, and TNI-cardiac. TNNI1 prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.

Sequence

Synthetic peptide located within the following region: MPEVERKPKITASRKLLLKSLMLAKAKECWEQEHEEREAEKVRYLAERIP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynn Yap et al.
NPJ Regenerative medicine, 8(1), 26-26 (2023-05-27)
Ischemic heart disease, which is often associated with irreversibly damaged heart muscle, is a major global health burden. Here, we report the potential of stem cell-derived committed cardiac progenitors (CCPs) have in regenerative cardiology. Human pluripotent embryonic stem cells were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service