Skip to Content
Merck
All Photos(1)

Key Documents

27668

Sigma-Aldrich

Collagenase Chromophore Substrate Test Substance

BioReagent, ≥95.0% (HPLC), powder or crystals, suitable for quantitative collagenase determination

Synonym(s):

4-Phenylazobenzyloxycarbonyl-Pro-Leu-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N4O5
CAS Number:
Molecular Weight:
466.53
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Collagenase Chromophore Substrate Test Substance, BioReagent, For quantitative collagenase determination

product line

BioReagent

Assay

≥95.0% (HPLC)

form

powder or crystals

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H]1CCCN1C(=O)OCc2ccc(cc2)\N=N\c3ccccc3)C(O)=O

InChI

1S/C25H30N4O5/c1-17(2)15-21(24(31)32)26-23(30)22-9-6-14-29(22)25(33)34-16-18-10-12-20(13-11-18)28-27-19-7-4-3-5-8-19/h3-5,7-8,10-13,17,21-22H,6,9,14-16H2,1-2H3,(H,26,30)(H,31,32)/b28-27+/t21-,22-/m0/s1

InChI key

GDWCICAVNWGMDQ-HCNWAXQFSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service