Skip to Content
Merck
All Photos(1)

Key Documents

Y0000039

Sodium ascorbate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+)-Sodium L-ascorbate, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NaO6
CAS Number:
Molecular Weight:
198.11
Beilstein:
3767246
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

sodium ascorbate

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

220 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].OC[C@H](O)[C@H]1OC(=O)C(O)=C1[O-]

InChI

1S/C6H8O6.Na/c7-1-2(8)5-3(9)4(10)6(11)12-5;/h2,5,7-10H,1H2;/q;+1/p-1/t2-,5+;/m0./s1

InChI key

PPASLZSBLFJQEF-RXSVEWSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium ascorbate may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

, Book author=
Handbook of Pharmaceutical Excipients (2009)
Sodium ascorbate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 6997-6998 (2019)
Marina A Kozuleva et al.
FEBS letters, 588(23), 4364-4368 (2014-10-15)
O2 reduction was investigated in photosystem I (PSI) complexes isolated from cyanobacteria Synechocystis sp. PCC 6803 wild type (WT) and menB mutant strain, which is unable to synthesize phylloquinone and contains plastoquinone at the quinone-binding site A1. PSI complexes from
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
Curt T Dellavalle et al.
International journal of cancer, 134(12), 2917-2926 (2013-11-19)
Nitrate and nitrite are precursors of endogenously formed N-nitroso compounds (NOC), known animal carcinogens. Nitrosation reactions forming NOCs can be inhibited by vitamin C and other antioxidants. We prospectively investigated the association between dietary nitrate and nitrite intake and risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service