Skip to Content
Merck
All Photos(1)

Key Documents

NIST1243

Nickel-chromium-cobalt alloy

NIST® SRM® 1243, UNS N07001 (disk form)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 1243 contains one disk, measuring ~ 34 mm in diameter and 19 mm thickness. For more information, please refer to the SDS and COA.
SRM 1243_cert

SRM 1243_SDS

Application

This Standard Reference Material (SRM) is intended for use in evaluating or calibrating spectrometric methods for analyzing Ni-Cr-Co alloys with similar compositions.

Features and Benefits

  • The NIST-certified mass fraction values for 14 constituents are reported on an as-received basis and are estimated using a Gaussian random effects model and the DerSimonian-Laird procedure.
  • Accompanied with Reference Mass Fraction Values for elements and Information Mass Fraction Value for tantalum.
  • Expiration details, instructions for handling, storage and use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Cobalt (Co),Chromium (Cr), Iron (Fe), Molybdenum (Mo), Niobium (Nb), Nickel (Ni), Silicon (Si), Titanium (Ti), Vanadium (V), Boron (B), Copper (Cu), Manganese (Mn), Phosphorus (P), Sulfur (S)

Reference/Informational value is provided for the following analytes.
Aluminium (Al), Carbon (C), Tungsten (W), Zirconium (Zr), Tantalum (Ta)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service