NIST1021
Glass beads - particle size distribution
NIST® SRM® 1021
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
beads
packaging
pkg of 4 g
manufacturer/tradename
NIST®
application(s)
glass & ceramic
Looking for similar products? Visit Product Comparison Guide
General description
SRM 1021_cert
SRM 1021_SDS
Other Notes
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service