Skip to Content
Merck
All Photos(1)

Key Documents

IRMM625

Chromium52Cr solution

IRMM®, certified reference material, 1 M in hydrochloric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

isotopic purity

 52Cr

manufacturer/tradename

JRC

concentration

1 M in hydrochloric acid

application(s)

general analytical

format

single component solution

storage temp.

−20°C

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM625

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service