Skip to Content
Merck
All Photos(1)

Documents

BCR505

Estuarine water (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Analysis Note

For more information please see:
BCR505

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service