Skip to Content
Merck
All Photos(1)

Documents

14549

Supelco

1-Methylpiperazine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2
CAS Number:
Molecular Weight:
100.16
Beilstein:
102724
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.466 (lit.)

bp

138 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CN1CCNCC1

InChI

1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3

InChI key

PVOAHINGSUIXLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methylpiperazine may be used as a solvent in an innovative blend in combination with piperazine and water in the ratio (30/10/60 w/w/w) for the post-combustion CO2 capture for power generation. This blend is further monitored for capture performance, degradation products and stability. 1-Methylpiperazine may also be used as a mimic template for the preparation of molecularly imprinted microspheres (MIMs) used for the extraction of four plant hormones such as indole-3-acetic acid (IAA), indole-3-propionic acid (IPA), indole-3- butyric acid (IBA) and 1-naphthaleneacetic acid (NAA) in banana samples, followed by analysis by high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

88.7 °F - closed cup

Flash Point(C)

31.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of 1-methylpiperazine.
Dorokhova, M. I., et al.
Pharmaceutical Chemistry Journal, 8.12, 737-740 (1974)
Monitoring of the blend 1-methylpiperazine/piperazine/water for post-combustion CO2 capture. Part 1: Identification and quantification of degradation products
Cuccia L, et al.
International Journal of Greenhouse Gas Control, 76, 215-224 (2018)
A simple and rapid procedure for phytate determination in soybeans and soy products.
Kwanyuen, Prachuab, and Joe W. Burton.
Journal of the American Oil Chemists' Society, 82.2, 81-85 (2005)
Hongyuan Yan et al.
The Analyst, 137(12), 2884-2890 (2012-05-04)
A highly selective molecularly imprinted solid-phase extraction (MISPE) combined with liquid chromatography-ultraviolet detection was developed for the simultaneous isolation and determination of four plant hormones including indole-3-acetic acid (IAA), indole-3-propionic acid (IPA), indole-3-butyric acid (IBA) and 1-naphthaleneacetic acid (NAA) in
H Welinder et al.
International archives of allergy and applied immunology, 79(3), 259-262 (1986-01-01)
The presence of specific IgE antibodies against conjugates between human serum albumin and piperazine, a compound previously known to cause occupational asthma, and N-methyl-piperazine, respectively, was demonstrated by RAST, RAST inhibition and skin prick tests in 2 asthmatic subjects occupationally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service