SYMBDTF01
Steritest® Symbio complete drain tray
For 2 canisters, FLEX pump, Low integration
Synonym(s):
Outlet
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151818
NACRES:
NB.79
material
316L stainless steel container (drain tray)
EPDM rubber seal
Agency
EP (2.6.1)
JP (4.06)
USP (71)
manufacturer/tradename
Steritest®
input
pharmaceutical(s)
application(s)
pharmaceutical
sterility testing
compatibility
for use with Steritest® Symbio pump
shipped in
ambient
General description
Steritest® Symbio complete drain tray includes stainless steel canister holder, plastic fluid collector, and seal. This tray is designed for 2 canisters FLEX Pump in low integration. The versatile Steritest® Symbio FLEX Pump can be installed in either an isolator or a laminar flow hood.
Application
Steritest® Symbio complete drain tray is a Steritest® Symbio FLEX Pump accessory for streamlining your workflow and increasing safety during sterility testing.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service