SCC717
3dGRO® Pancreatic Organoids (OCIP.180)
Synonym(s):
organoid, PDO, cancer organoid, tumor organoid, 3d cell culture, pancreatic cancer, PDCAC organoid, cancer, organoid cell line
About This Item
biological source
human
Quality Level
packaging
vial of ≥1500 organoids/vial vials
manufacturer/tradename
Millipore
technique(s)
cell culture | mammalian: suitable
cell culture | stem cell: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
General description
Recent breakthroughs in three-dimensional (3D) culture methods have led to the development of organoid culture platforms, which allow researchers to perform translational research on long-term in vitro cultures not limited by barriers present in 2D culture or animal models. We now offer a comprehensive collection of patient derived pancreatic ductal adenocarcinoma cancer (PDAC) organoids for cancer research and drug discovery applications.
Cell Line Characteristics
- Sex: Male
- Age: 64 Years
- Organ: Pancreas (Xenograph from Primary Tumor)
- Disease: Ductal Adenocarcinoma (KRAS mutation: Wild-type)
Application
- Viability: >=1500 viable organoids/vial
- Organoid Growth: Pass
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Mycoplasma Contamination: Negative
- STR Profile: Pass
Features and Benefits
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service