PCC030C30E
Pellicon® Capsule with Ultracel® Membrane
30 kDa, C Screen, 3 m2 manifold extender with AseptiQuik® L connectors
About This Item
Recommended Products
material
EPDM rubber seal
PPO/PS blend (Core)
PPO/PS blend housing
TPE seal
composite regenerated cellulose (CRC) membrane
epoxy adhesive
polyester screen
polypropylene screen
polyurethane adhesive
Quality Level
sterility
sterile; γ-irradiated
product line
Pellicon®
storage condition
room temperature (15-30°C)
parameter
4-8 L/min-m2 flow rate
50 psi pressure (3.5 bar) at 4-30 °C (Forward Transmembrane Pressure (TMP))
60 psi max. pressure (4.1 bar) at 4-30 °C (Maximum Inlet Pressure)
technique(s)
ultrafiltration: suitable
H
46.9 cm (18.5 in.)
L
45.9 cm (18.1 in.)
filtration area
3 m2
screen size
, Type C screen (coarse screen)
size
48.2 cm (19.0 in.)
volume
1288 mL , in feed channel
1627 mL , in permeate channel
impurities
≤5 ppm TOC ( in retentate effluent, after a WFI flush of 20 L/m²)
matrix
Ultracel®
pore size
30 kDa
pH range
2-13
fitting
inlet/outlet connection (Feed: AseptiQuik® L Connector)
inlet/outlet connection (Permeate: AseptiQuik® L Connector)
inlet/outlet connection (Retentate: AseptiQuik® L Connector)
General description
Application
Features and Benefits
Gamma Sterilized and preseravtive-free - ready to process in minutes
True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families
Packaging
Other Notes
Comprehensive documentation to support qualification, risk assessment and process optimization needs.
Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
Convenient 24/7 access to up-to-date product information.
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service