Skip to Content
Merck
All Photos(1)

Key Documents

MABT1548

Sigma-Aldrich

Anti-CD97 Antibody, clone CLB‐CD97/1

Synonym(s):

Adhesion G protein-coupled receptor E5, Leukocyte antigen CD97

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
CLB?CD97/1, monoclonal
application:
ELISA
FACS
IF
IHC
IP
inhibition assay
species reactivity:
human
technique(s):
ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CLB?CD97/1, monoclonal

mol wt

calculated mol wt 92 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable

isotype

IgG2a

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... ADGRE5(976)

Specificity

Clone CLB‐CD97/1 is a mouse monoclonal antibody that detects CD97 (Adhesion G protein-coupled receptor E5).

Immunogen

NIH3T3 cells stably expressing human CD97.

Application

Quality Control Testing

Evaluated by Flow Cytometry in THP-1 cells.

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD97 in one million THP-1 cells.

Tested Applications

Flow Cytometry Analysis: A representative lot detected CD97 in Flow Cytometry applications (Hamann, J., et al. (1996). 184(3):1185-9; Kwakkenbos, M.J., et al. (2002). J Leukoc Biol. 71(5):854-62).

Inhibition Assay: A representative lot of this antibody inhibited erythrocyte adhesion to CD97-transfected COS cells.(Hamann, J., et al. (1996). 184(3):1185-9; Hamann, J., et al. (1998). Eur J Immunol.;28(5):1701-7).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CD97 in Immunoprecipitation applications (Hamann, J., et al. (1996). 184(3):1185-9).

Immunohistochemistry Applications: A representative lot detected CD97 in Immunohistochemistry applications (Hamann, J., et al. (1999). Arthritis Rheum. 42(4):650-8; Steinert, M., et al. (2002). Am J Pathol. 161(5):1657-67).

Immunofluorescence Analysis: A representative lot detected CD97 in Immunofluorescence applications (Hamann, J., et al. (1998). Eur J Immunol. 28(5):1701-7; Hamann, J., et al. (1999). Arthritis Rheum. 42(4):650-8; Steinert, M., et al. (2002). Am J Pathol. 161(5):1657-67).

ELISA Analysis: A representative lot detected CD97 in ELISA applications (Hamann, J., et al. (1999). Arthritis Rheum. 42(4):650-8; Steinert, M., et al. (2002). Am J Pathol. 161(5):1657-67).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Adhesion G protein-coupled receptor E5 (UniProt: P48960; also known as Leukocyte antigen CD97, CD97) is encoded by the ADGRE5 (also known as CD97) gene (Gene ID: 976) in human. CD97 is a multi-pass, homodimeric, membrane glycoprotein of the G-protein coupled receptor 2 family that is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to produce the mature protein that contains four extracellular domains, seven transmembrane domains, and four cytoplasmic domains. The extracellular domain contains five EGF-like domains. The second and third EGF-like domains contain calcium binding sites, which are important in certain ligand interactions that are calcium-dependent. The fourth EGF-like domain mediates binding of chondroitin sulfate. The extracellular domain also contains one Arg-Gly-Asp (RGD) motif that serves as a recognition site for integrin binding. CD97 levels are quickly upregulated upon activation of most leukocytes. It is involved in cell adhesion and migration through interactions with cell surface proteins and components of the extracellular matrix. Three ligands for CD97 have been recognized: CD55 that serves as a negative regulator of complement cascade; chondroitin sulfate, and integrin 5β1. CD97 is broadly expressed and is found on most hematopoietic cells, including activated lymphocytes, monocytes, macrophages, dendritic cells, and granulocytes. Its expression is also observed on smooth muscle cells. Higher expression of CD97 is reported in thyroid, colorectal, esophageal, and pancreatic carcinomas. Its expression levels correlate with dedifferentiation, migration, and invasion in tumor cells. Three isoforms of CD97 have been described that are produced by alternative splicing. (Ref.: Safaee, M., et al. (2013). Int. J. Oncology. 43(5); 1343-1350; Steinert, M., et al. (2002). Am. J. Pathol. 161(5); 1657-1667; Kwakkenbos, MJ., et al. (2002). J. leukocyte Biol. 71(5); 854-862; Hamann, J., et al. (1996). J. Exp. Med. 184(3); 1185-1189).

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without preservatives.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service