Skip to Content
Merck
All Photos(2)

Key Documents

MABF299

Sigma-Aldrich

Anti-CCR2 Antibody, clone 8C3.1

clone 8C3.1, from mouse

Synonym(s):

C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CCR-2, CCR2, CD192, MCP-1-R, Monocyte chemoattractant protein 1 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8C3.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CCR2 (729230)

Related Categories

General description

C-C chemokine receptor type 2 (UniProt P41597; also known as C-C CKR-2, CC-CKR-2, CCR-2, CCR2, CD192, MCP-1-R, Monocyte chemoattractant protein 1 receptor) is encoded by the CCR2 (also known as CMKBR2) gene (Gene ID 729230) in human. CC chemokine receptors (CCRs) are 7-transmembrane G protein-coupled receptors (GPCRs) that bind CC chemokines (CCLs) and mediate their biological activities. CCRs and CCLs play a central role in physiological and pathological recruitment of immune cells. CCR2 can interact with CCL2, CCL8 and CCL16, it is found on the surface of monocytes, activated memory T cells, B cells, and basophils in humans, and also in peritoneal macrophages in mice. CCR2 plays an important role in the recruitment of monocytes/macrophages and T cells. CCR2-knockout mice show a partial defect in macrophage function, decreased Th1 immune response, reduced delayed-type hypersensitivity reaction, and are more susceptible to pathogenic bacteria infections. Human CCR2 is a 7-transmembrane (a.a. 43-70, 81-100, 115-136, 154-178, 207-226, 244-268, 286-309) GPCR with an N-terminal extracellular domain (a.a. 1-42) and a C-terminoal cytoplasmic domain (a.a. 310-374).

Specificity

Clone 8C3.1 targets the N-terminal extracelluar domain of human CCR2 spliced isoforms 1&2 reported by Uniprot (P41597).

Immunogen

GST-tagged recombinant human N-terminal extracellular domain.

Application

Flow Cytometry Analysis: 2 µL from a representative lot detected surface CCR2 expression among the gated lymphocytes population from one million human PMBCs.
Research Category
Inflammation & Immunology
This mouse monoclonal Anti-CCR2 Antibody, clone 8C3.1, Cat. No. MABF299 is validated for use in Flow Cytometry and Western Blotting for the detection of CCR2.

Quality

Evaluated by Western Blotting in U937 cell lysate.

Western Blotting Analysis (WB): A 100,000 dilution of this antibody detected CCR2 in 10 µg of human lymphoma U937 monocyte lysate.

Target description

~40-50 kDa observed. 41.92 kDa (isoform 1) and 41.06 kDa (isoform 2) calculated. Larger-than-calculated sizes and smeary banding pattern are consistent with the detection of differentially glycosylated CCR2 species. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service