Skip to Content
Merck
All Photos(2)

Key Documents

MABE270

Sigma-Aldrich

Anti-phospho STAT6 (Tyr641) Antibody, clone 19E2.1

clone 19E2.1, from mouse

Synonym(s):

Signal transducer and activator of transcription 6, IL-4 Stat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

19E2.1, monoclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr641)

Gene Information

human ... STAT6(6778)

Related Categories

General description

Signal transducer and activator of transcription 6 (STAT6) transduces signals from extracellular cytokines during adaptive immunity and has recently been found to be important in the innate immune response to viral infection. STAT6 is also involved in the attenuation of the Th1 and Th17 in the nephritogenic immune response. The consequence of STAT6 activity has been associated with the pathogenesis of autoimmune disease and has been found to be actively imported to the nucleus.

Specificity

This antibody recognizes STAT6 phosphorylated at (Tyr641).

Immunogen

Epitope: Phosphorylated (Tyr641)
KLH-conjugated linear peptide corresponding to human STAT6 phosphorylated at (Tyr641).

Application

Anti-phospho STAT6 (Tyr641) Antibody, clone 19E2.1 is a mouse monoclonal antibody for detection of phospho STAT6 (Tyr641) also known as Signal transducer & activator of transcription 6, IL-4 Stat & has been validated in WB, FC.
Flow Cytometry Analysis: 2 µg/mL from a representative lot detected STAT6 in human peripheral blood leukocytes.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in NIH/3T3 cell lysates treated and untreated with PDGF.

Western Blot Analysis: 0.5 µg/mL of this antibody detected STAT6 on 10 µg of PDGF untreated and treated NIH/3T3 cell lysates.

Target description

~165 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysates treated and untreated with PDGF

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service