Skip to Content
Merck
All Photos(1)

Key Documents

MAB3478

Sigma-Aldrich

Anti-1-cys Peroxiredoxin Antibody, clone 8H11

ascites fluid, clone 8H11, Chemicon®

Synonym(s):

aiPLA2, Peroxiredoxin 6, Non-selenium Glutathione Peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
8H11, monoclonal
application:
ICC
IHC
WB
species reactivity:
human, rat
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8H11, monoclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PRX(57716)

General description

1-Cys-Peroxiredoxin is a cytosolic protein that is involved in the regulation of cellular redox potential. 1-Cys peroxiredoxin is a bifunctional enzyme with glutathione peroxidase and phospholipase A2 activities (Chen, 2000). It can reduce hydrogen peroxide, short chain fatty acids, and phospholipid hydroperoxides and is believed to play a role in phospholipid turnover as well as in the protection against oxidative injury.

Specificity

1-Cys-Peroxiredoxin (Prx VI).

Immunogen

Recombinant human 1-Cys peroxiredoxin.

Application

Detect 1-cys Peroxiredoxin using this Anti-1-cys Peroxiredoxin Antibody, clone 8H11 validated for use in WB, IC, IH.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress
Western blot on rat lung lysate (24 kDa) or recombinant human protein.

Immunohistochemistry on 4% paraformaldehyde fixed rat lung tissue.

Immunocytochemistry on rat type II alveolar epithelial cells.

Optimal working dilutions must be determined by the end user.

Physical form

Ascites fluid. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seunghyi Kook et al.
The Journal of biological chemistry, 291(16), 8414-8427 (2016-02-26)
The Hermansky Pudlak syndromes (HPS) constitute a family of disorders characterized by oculocutaneous albinism and bleeding diathesis, often associated with lethal lung fibrosis. HPS results from mutations in genes of membrane trafficking complexes that facilitate delivery of cargo to lysosome-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service