HTS068M
ChemiSCREEN Membrane Preparation Recombinant Human APJ Apelin Receptor
Human APJ GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... APLNR(187)
General description
Application
Biochem/physiol Actions
Quality
10 µg/well | 5 µg/well | |
---|---|---|
Signal:Background | 33 | 50 |
Specific Binding (cpm) | 17297 | 15015 |
SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax for [125I]-Apelin binding: 3.85 pmol/mg protein
Kd for [125I]-Apelin binding: ~0.4 nM
Specifications
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]-Apelin. (Perkin Elmer#:NEX-393 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 20-fold signal:background with 125I labeled Apelin at 0.25 nM
Physical form
Packaging method: Membranes protein were adjusted to the indicated concentration in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service