Skip to Content
Merck
All Photos(1)

Documents

ABN1737

Sigma-Aldrich

Anti-BHLHE40/DEC1 Antibody

serum, from rabbit

Synonym(s):

Class E basic helix-loop-helix protein 40, bHLHe40, Class B basic helix-loop-helix protein 2, bHLHb2, Differentially expressed in chondrocytes protein 1, DEC1, Enhancer-of-split and hairy-related protein 2, SHARP-2, Stimulated by retinoic acid gene 13 pr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), rat (based on 100% sequence homology), horse (based on 100% sequence homology), canine (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Class E basic helix-loop-helix protein 40 (UniProt O14503; also known as Basic helix-loop-helix domain containing class B2, Class B basic helix-loop-helix protein 2, Differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, Differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP-2, Stimulated by retinoic acid gene 13 protein) is encoded by the BHLHE40 (also known as BHLHB2, DEC1, HLHB2, SHARP2, STRA13) gene (Gene ID 8553) in human. DEC1 (differentially expressed in chondrocytes 1) and DEC2 are E-box-binding transcription factors that exhibit a circadian expression pattern. Both proteins act to repress the Clock/Bmal1-activated E-box promoters via direct E-box binding rather than Bmal1 interaction. In addition, DEC1 is shown to negatively regulate the expression of DEC2. Markedly increased DEC1 expression is observed in colon carcinomas, whereas the opposite trend is seen with DEC2. Likewise cellular DEC1 over-expression results in a proportional decrease in DEC2 level.

Immunogen

Epitope: N-terminal
KLH-conjugated linear peptide corresponding to human BHLHE40/DEC1.

Application

Anti-BHLHE40/DEC1 Antibody is an antibody against BHLHE40/DEC1 for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in K562 nuclear extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected BHLHE40/DEC1 in 10 µg of K562 nuclear extract.

Target description

~48 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service