Skip to Content
Merck
All Photos(1)

Key Documents

8.51039

Sigma-Aldrich

Boc-NH-(PEG)-COOH . DCHA (9 atoms)

Novabiochem®

Synonym(s):

Boc-NH-(PEG)-COOH . DCHA (9 atoms), Boc-8-amino-3,6-dioxaoctanoic acid · DCHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO6 · xC12H23N
CAS Number:
Molecular Weight:
263.29
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥99% (TLC)

form

powder

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

[N+H2](C2CCCCC2)C1CCCCC1.N(CCOCCOCC(=O)[O-])C(=O)OC(C)(C)C

InChI

1S/C12H23N.C11H21NO6/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-11(2,3)18-10(15)12-4-5-16-6-7-17-8-9(13)14/h11-13H,1-10H2;4-8H2,1-3H3,(H,12,15)(H,13,14)

InChI key

SWUXEYKTUQROOO-UHFFFAOYSA-N

General description

9-Atom long aminoacid PEG spacer. Can be introduced using Boc SPPS protocols

Linkage

Replaces: 01-63-0205

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (LC-MS): 262.1 - 264.1
Purity (TLC(011A)): ≥ 99 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service