Skip to Content
Merck
All Photos(1)

Key Documents

8.21814

Sigma-Aldrich

4-Hydroxybenzoic acid

for synthesis

Synonym(s):

4-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(OH)C6H4COOH
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-804-9
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

autoignition temp.

>250 °C

potency

>5000 mg/kg LD50, oral (Rat)

pH

3.3 (20 °C, 1 g/L in H2O)

mp

213-216 °C

solubility

5 g/L

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

Oc1ccc(cc1)C(=O)O

InChI

1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

FJKROLUGYXJWQN-UHFFFAOYSA-N

Application

  • Production and Applications of Hydroxy Acids: This study discusses the synthesis of hydroxy acids including 4-hydroxybenzoic acid, focusing on enzymatic routes which are pivotal for drug discovery due to their application in creating more efficient and environmentally friendly production methods (Bhalla et al., 2014).
  • Microbial Synthesis from Renewable Feedstocks: Highlights microbial production methods for 4-hydroxybenzoic acid from renewable feedstocks, crucial for sustainable chemical synthesis in pharmaceuticals and material science (Chen et al., 2021).
  • Laccase-mediated Oligomerization: Explores the enzymatic oligomerization of 4-hydroxybenzoic acid which could lead to new materials with improved stability and functionality, useful in material science and drug delivery systems (Slagman et al., 2016).
  • Antiviral Properties: Discusses the potential of 4-hydroxybenzoic acid derived from the autooxidation of catechinic acid as an antiviral agent against Herpes Simplex Virus, offering potential applications in drug development (Alfei et al., 2022).
  • Soil Microbial Community Impact: Investigates the effect of 4-hydroxybenzoic acid on grape soil microbial communities, emphasizing its potential environmental impact and applications in sustainable agriculture and academic studies (Guo et al., 2015).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 214 °C
Melting range (upper value): ≤ 217 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service