Skip to Content
Merck
All Photos(1)

Key Documents

8.20259

Sigma-Aldrich

1,4-Dihydroxyanthraquinone

for synthesis

Synonym(s):

1,4-Dihydroxyanthraquinone, Quinizarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-368-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.3 hPa ( 197 °C)

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

potency

>5000 mg/kg LD50, oral (Rat)

bp

450 °C/1013 hPa

mp

195-198 °C

transition temp

flash point 222 °C

bulk density

350 kg/m3

storage temp.

2-30°C

SMILES string

Oc1c2c(c(cc1)O)C(=O)c3c(cccc3)C2=O

InChI

1S/C14H8O4/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6,15-16H

InChI key

GUEIZVNYDFNHJU-UHFFFAOYSA-N

Application

1,4-Dihydroxyanthraquinone for synthesis. CAS 81-64-1, molar mass 240.21 g/mol.

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Melting range (lower value): ≥ 195 °C
Melting range (upper value): ≤ 198 °C
Identity (IR): passes test

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

431.6 °F

Flash Point(C)

222 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service