Skip to Content
Merck
All Photos(1)

Key Documents

8.18255

Sigma-Aldrich

Paraldehyde

for synthesis

Synonym(s):

Paraldehyde, 2,4,6-Trimethyl-1,3,5-trioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-639-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

13.3 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

235 °C

potency

2711 mg/kg LD50, oral (Rat)
13959 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-17.0 % (v/v)

bp

124 °C/1013 hPa

mp

12 °C

transition temp

flash point 27 °C

solubility

120 g/L

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(OC(OC1C)C)C

InChI

1S/C6H12O3/c1-4-7-5(2)9-6(3)8-4/h4-6H,1-3H3

InChI key

SQYNKIJPMDEDEG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.992 - 0.994
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service