Skip to Content
Merck
All Photos(1)

Key Documents

8.00945

Sigma-Aldrich

2-Chloroethanol

for synthesis

Synonym(s):

Ethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.22
form:
liquid
Assay:
≥99.0% (GC)

vapor pressure

6.6 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

425 °C

expl. lim.

5-16 % (v/v)

pH

6-7 (20 °C, 500 g/L in H2O)

kinematic viscosity

3.43 cSt(23 °C)

bp

129 °C/1013 hPa

mp

-63 °C

transition temp

flash point 55 °C

density

1.2 g/cm3 at 20 °C

General description

for synthesis

Application

2-Chloroethanol for synthesis. CAS 107-07-3, EC Number 203-459-7, molar mass 80.51 g/mol.

Storage and Stability

Store at +5°C to +30°C.

Analysis Note

Assay (GC, area%):≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C):1.201 - 1.203
Water (according to Karl Fischer):≤ 0.10 %
Identity (IR):passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service