648463
Triton™ X-100 Solution
10% in H₂O, PROTEIN GRADE®
Synonym(s):
Triton™ X-100, Polyethylene Glycol- p-isooctylphenyl Ether, Octylphenoxypolyethoxyethanol, Polyethylene Glycol-p-isooctylphenyl Ether, Octylphenoxypolyethoxyethanol, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether
About This Item
Recommended Products
Product Name
Triton™ X-100, PROTEIN GRADE® Detergent, 10% Solution, Sterile-Filtered, Non-ionic detergent useful for isolation of membrane proteins.
description
Merck USA index - 14, 6761
Quality Level
form
liquid
mol wt
micellar avg mol wt 80,000
average mol wt 625
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
aggregation number
100-155
impurities
<200 μM Aldehyde
<200 μM peroxide
color
clear
conductivity
<50 μmho
CMC
0.2-0.9 mM (20-25°C)
transition temp
cloud point 65 °C
pour point ~7 °C
solubility
water: soluble
HLB
13.5
shipped in
ambient
storage temp.
2-8°C
SMILES string
CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1
InChI
1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3
InChI key
HNLXNOZHXNSSPN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Biochem/physiol Actions
Packaging
Warning
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.
Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.
Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.
Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service