Skip to Content
Merck
All Photos(1)

Key Documents

539053-5T

Millipore

Protein Molecular Weight Markers, HPLC

Synonym(s):

Molecular weight standards, Protein markers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105337
NACRES:
NC.01
Pricing and availability is not currently available.

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

Contains five highly purified proteins ranging in molecular weight from 12.4 kDa to 290 kDa. Formulated to give peaks of approximately the same intensity for each protein when run on a TSK-Gel G300SW* column after reconstituting with elution buffer and monitored at 280 nm.



Each vial contains sufficient material for approximately 5–10 tests. There is 200 µg of each protein per vial x 5 proteins = 1000 µg total protein per vial. Each vial also contains ~5 mg sucrose as a carrier.



*Toyosoda Industry Co., Ltd.
Note: 5T = 5 Tests.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute with 100 µl of 100 mM sodium phosphate buffer, pH 7.0 containing 200 mM NaCl. If the material is reconstituted with lower ionic strength buffer (i.e., <0.1 M NaCl) the cytochrome c may interact nonspecifically with the silica gel column and migrate incorrectly. Apply 20 µl to an analytical grade column such as TSK-Gel G3000SW (0.75 x 60 cm).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service