Skip to Content
Merck
All Photos(1)

Key Documents

524636

Sigma-Aldrich

Phosphatase Inhibitor Cocktail II

lyophilized solid, for the inhibition of acid and alkaline phosphatases and protein tyrosine phosphatases, This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Inhibitor Cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54
form:
solid
solubility:
water: soluble
storage temp.:
2-8°C

Product Name

Phosphatase Inhibitor Cocktail Set II, Lyophilized, The Phosphatase Inhibitor Cocktail Set II, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts, including samples containing detergents. Provided as a set of five vials. Each vial, when reconstituted with 1 ml H2O, contains Imidazole (200 mM), Sodium Fluoride (100 mM), Sodium Molybdate (115 mM), activated Sodium Orthovanadate (100 mM), and Sodium Tartrate Dihydrate (400 mM). Reconstitute each vial with 1 ml H2O.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Dilute 1:100 just prior to use.
Reconstitute each vial with 1 ml H₂O.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jrhau Lung et al.
American journal of translational research, 13(10), 11194-11208 (2021-11-18)
Cullin 4A (Cul4A) reportedly has oncogenic roles in several cancer types by regulating tumor suppressors through the ubiquitination and proteolysis of the tumor suppressor. In addition, Cul4A is associated with chemosensitivity to chemotherapy drugs. This study investigated the association between
Delphine Séhédic et al.
Frontiers in bioengineering and biotechnology, 8, 602998-602998 (2021-03-16)
Inhibition of the PI3K/Akt/mTOR signaling pathway represents a potential issue for the treatment of cancer, including glioblastoma. As such, rapamycin that inhibits the mechanistic target of rapamycin (mTOR), the downstream effector of this signaling pathway, is of great interest. However
Jorge Beleza et al.
International journal of molecular sciences, 23(7) (2022-04-13)
Mothers' antenatal strategies to improve the intrauterine environment can positively decrease pregnancy-derived intercurrences. By challenging the mother-fetus unit, gestational exercise (GE) favorably modulates deleterious stimuli, such as high-fat, high-sucrose (HFHS) diet-induced adverse consequences for offspring. We aimed to analyze whether
Zeda Zhang et al.
Cancer cell, 38(2), 279-296 (2020-07-18)
Despite the development of second-generation antiandrogens, acquired resistance to hormone therapy remains a major challenge in treating advanced prostate cancer. We find that cancer-associated fibroblasts (CAFs) can promote antiandrogen resistance in mouse models and in prostate organoid cultures. We identify
Jelena Stevanović-Silva et al.
International journal of environmental research and public health, 20(2) (2023-01-22)
Maternal high-caloric nutrition and related gestational diabetes mellitus (GDM) are relevant modulators of the intrauterine environment, increasing the risk of liver metabolic alterations in mothers and offspring. In contrast, as a non-pharmacological approach against metabolic disorders, exercise is highly recommended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service