Skip to Content
Merck
All Photos(1)

Key Documents

427572

Sigma-Aldrich

Bovine Lactosyl Ceramide

≥98% (TLC), solid, glycosphingolipid precursor, Calbiochem®

Synonym(s):

Lactosyl Ceramide, Bovine, Galβ,1,4Glc-Cer, LacCer, LC, LC, Galβ,1,4Glc-Cer, LacCer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H101NO13
CAS Number:
Molecular Weight:
960.37
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Product Name

Lactosyl Ceramide, Bovine, A neutral glycosphingolipid (GSL) that is the common precursor of the ganglio-, globo-, globoiso-, lacto-, neolacto-, and muco-series of GSL.

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless

solubility

ethanol: 1 mM
methanol: 1 mM

shipped in

ambient

storage temp.

−20°C

SMILES string

N(C(C(O)\C=C\CCCCCCCCCCCCC)CO[C@@H]1O[C@@H]([C@@H]([C@@H]([C@H]1O)O)OC2O[C@@H]([C@@H]([C@@H]([C@H]2O)O)O)CO)CO)C(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C48H91NO13/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(53)49-36(37(52)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2)35-59-47-45(58)43(56)46(39(34-51)61-47)62-48-44(57)42(55)41(54)38(33-50)60-48/h29,31,36-39,41-48,50-52,54-58H,3-28,30,32-35H2,1-2H3,(H,49,53)/b31-29+/t36?,37?,38-,39-,41+,42+,43-,44-,45-,46+,47-,48?/m1/s1

InChI key

VOZHMDQUIRUFQW-AIRKALMMSA-N

General description

A neutral glycosphingolipid (GSL) that is the common precursor of the ganglio-, globo-, globoiso-, lacto-, neolacto-, and muco-series of GSL. Promotes a time- and concentration-dependent proliferation of aortic smooth muscle cells via specific activation of p44 MAP kinase. Also stimulates Ras-GTP loading, MEK and Raf kinases, p44 MAP kinase, and c-fos expression in human aortic smooth muscle cells.
A neutral glycosphingolipid (GSL) which is the common precursor of the ganglio-, globo-, globoiso-, lacto-, neolacto-, and muco- series of GSL. Promotes a time- and concentration-dependent proliferation of aortic smooth muscle cells via specific activation of p44 MAP kinase. Also stimulates Ras-GTP loading, MEK and Raf kinases, p44 MAP kinase and c-fos expression in human aortic smooth muscle cells.



Molecular weight: 960.4 (based on tricosanyl -23:0)
M.W. 960.4 (based on tricosanoyl - 23:0).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A neutral glycosphingolipid (GSL) that is the common precursor of the ganglio-, globo-, globoiso-, lacto-, neolacto-, and muco-series of GSL
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Bhunia, A.K., et al. 1997. J. Biol. Chem.272, 15642.
Bhunia, A.K., et al. 1996. J. Biol. Chem.271, 10660.
Zacharias, C., et al. 1994. J. Biol. Chem.269, 13313.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service