Skip to Content
Merck
All Photos(1)

Key Documents

236005

Sigma-Aldrich

COX-1 Inhibitor, FR122047

The COX-1 Inhibitor, FR122047, also referenced under CAS 130717-51-0, controls the biological activity of COX-1. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

COX-1 Inhibitor, FR122047, FR 122047, 1-[(4,5-Bis(4-methoxyphenyl-2-thiazoyl)carbonyl]-4-methylpiperazine, Hydrochloride, 4,5-Bis(4-methoxyphenyl-2-[(1-methylpiperazin-4-yl)carbonyl]thiazole, Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N3O3S · xHCl
CAS Number:
Molecular Weight:
423.53 (free base basis)
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale yellow

solubility

1 M HCl: 2 mg/mL
water: 4 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1c4ccc(cc4)OC)c3ccc(cc3)OC)C(=O)N2CCN(CC2)C.[Cl-].[H+]

InChI

1S/C23H25N3O3S.ClH/c1-25-12-14-26(15-13-25)23(27)22-24-20(16-4-8-18(28-2)9-5-16)21(30-22)17-6-10-19(29-3)11-7-17;/h4-11H,12-15H2,1-3H3;1H

InChI key

YWMAVHIKOAOSFM-UHFFFAOYSA-N

General description

A cell-permeable trisubstituted thiazole compound that acts as a potent and selective inhibitor of COX-1 (IC50 = 28 nM and 65 µM for human recombinant COX-1 and COX-2, respectively). Displays ~2,300 fold greater selectivity towards inhibition of COX-1 over COX-2. Reported to be orally active and display analgesic, anti-platelet and anti-inflammatory properties.
A potent, cell-permeable, and selective inhibitor of COX-1 (IC50 = 28 nM; human recombinant COX-1). Does not significantly inhibit COX-2 (IC50 = 65 µM, human recombinant COX-2). Reported to be orally active and displays analgesic properties in chemical nociceptive animal models. Also reported to exhibit anti-inflammatory effects and suppress prostaglandin E2 and thromboxane 2 levels in collagen-induced arthritis in rats. Shown to be 100 times more potent than aspirin in inhibiting arachidonic acid- and collagen-induced human and guinea pig platelet aggregation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
COX-1, COX-2
Product does not compete with ATP.
Reversible: no
Target IC50: 28 nM and 65 µM for human recombinant COX-1 and COX-2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ochi, T., and Goto, T. 2002. Br. J. Pharmacol.135, 782.
Ochi, T., et al. 2000. Eur. J. Pharmacol.391, 49.
Tanaka, A., et al. 1994. J. Med. Chem.37, 1189.
Dohi, M., et al. 1993. Eur. J. Pharmacol.243, 179.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service