Skip to Content
Merck
All Photos(1)

Key Documents

189406

Sigma-Aldrich

Aurora Kinase/Cdk Inhibitor

The Aurora Kinase/Cdk Inhibitor, also referenced under CAS 443797-96-4, controls the biological activity of Aurora Kinase/Cdk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Aurora Kinase/Cdk Inhibitor, 4-(5-Amino-1-(2,6-difluorobenzoyl)-1H-[1,2,4]triazol-3-ylamino)-benzenesulfonamide, JNJ-7706621, Aurora Kinase Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12F2N6O3S · xC2H3N
CAS Number:
Molecular Weight:
394.36
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 15 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(c(ccc1)F)C(=O)N2NC(=NC2=N)Nc3ccc(cc3)[S](=O)(=O)N

InChI

1S/C15H12F2N6O3S/c16-10-2-1-3-11(17)12(10)13(24)23-14(18)21-15(22-23)20-8-4-6-9(7-5-8)27(19,25)26/h1-7H,(H2,19,25,26)(H3,18,20,21,22)

InChI key

KDKUVYLMPJIGKA-UHFFFAOYSA-N

General description

A cell-permeable triazolylsulfonamido compound that acts a reversible, ATP-competitive kinase inhibitor with selectivities towards Aurora kinases (IC50 = 11 and 15 nM for Aurora-A, Aurora-B, respectively) and Cdk′s (IC50 = 9, 4, and 3 nM for Cdk1/B, Cdk2/A, and Cdk2/E, respectively). Displays antitumor properties both in vitro (IC50 in the range of 112 - 514 nM against various human cancer cell lines) and in a xenograft murine model in vivo (75-100 mg/kg, i.p.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Aurora Kinase/Cdk
Product competes with ATP.
Reversible: yes
Target IC50: 11 and 15 nM for Aurora-A, Aurora-B, respectively; 9, 4, and 3 nM for Cdk1/B, Cdk2/A, and Cdk2/E, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Emanuel, S., et al. 2005. Cancer Res.65, 9038.
Lin R., et al. 2005. J. Med. Chem.48, 4208.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service