Skip to Content
Merck
All Photos(2)

Key Documents

1.09512

Supelco

Potassium iodide-starch paper

roll L 4.8 m , with color scale, reag. Ph. Eur., MQuant®

Synonym(s):

Potassium iodide-starch chemical test paper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

Agency

reag. Ph. Eur.

Quality Level

description

semi-quantitative

product line

MQuant®

reaction suitability

reagent type: oxidant

roll L

4.8 m , with color scale

detection method

colorimetric

storage temp.

15-25°C

General description

Potassium iodine paper is used for the determination of oxidizing agents. In contact with oxidizing agents such as sulfite or sulfur dioxide, it turns blue. This is a qualitative test paper, telling whether oxidizing agents are in the sample or not. For a semiquantitative or quantitative result, please consider a test for a specific oxidizing agent from our MQuant®, Reflectoquant® or Spectroquant® portfolio.

Analysis Note

Application test: conforms

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service