Skip to Content
Merck
All Photos(1)

Key Documents

1.07838

Supelco

Antimony(III) chloride

for analysis EMSURE® ACS

Synonym(s):

Antimony(III) chloride, Antimonous acid, Antimony trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl3Sb
CAS Number:
Molecular Weight:
228.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-047-2
NACRES:
NA.21
Assay:
≥99.0% (bromometric)
grade:
ACS reagent
form:
solid
solubility:
931 g/L (Hydrolysis)

grade

ACS reagent

Quality Level

vapor pressure

0.16 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.0% (bromometric)

form

solid

impurities

≤0.005% Substances insoluble in hydrochloric acid
≤0.02% Matter insoluble in chloroform

bp

233 °C/1013 hPa

mp

73 °C

solubility

931 g/L (Hydrolysis)

density

3.14 g/cm3 at 20 °C

bulk density

1600 kg/m3

anion traces

sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.002%
K: ≤0.01%
Na: ≤0.005%
Pb: ≤0.003%

storage temp.

2-30°C

InChI

1S/C22H23ClN2O8.3ClH.Sb/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;;;;/h4-5,7-8,15,26-27,31-33H,6,24H2,1-3H3;3*1H;/q;;;;+3/p-3/b20-13+;;;;/t7-,8-,15-,21-,22-;;;;/m0..../s1

InChI key

LIARQHHWPMTMFO-CRPMINNHSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Syntheses and characterization of novel antimony (III) and bismuth (III) derivatives containing β-enamino ester along with antimicrobial evaluation, DFT calculation, and cytotoxic study.: This study involved the synthesis and characterization of novel antimony(III) chloride derivatives. The research highlighted its applications in antimicrobial evaluation, density functional theory (DFT) calculations, and cytotoxic studies, demonstrating the compound′s potential in developing new antimicrobial agents (Sangwan et al., 2024).

Linkage

Replaces: AX1675

Analysis Note

Assay (bromometric): ≥ 99.0 %
Matter insoluble in chloroform: ≤ 0.02 %
Substances insoluble in hydrochloric acid: ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
As (Arsenic): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.01 %
Na (Sodium): ≤ 0.005 %
Pb (Lead): ≤ 0.003 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service