Skip to Content
Merck
All Photos(2)

Documents

1.06498

Supelco

Sodium hydroxide

pellets for analysis EMSURE®

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
EC Index Number:
215-185-5
NACRES:
NA.21

Quality Level

Agency

suitable for SM 4500 - NH3

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

EMSURE®

Assay

≥99% NaOH basis (acidimetric)

form

pellets

impurities

≤0.0003% Total nitrogen (N)

pH

>14 (20 °C, 100 g/L in H2O)

mp

318 °C (lit.)

solubility

1090 g/L

density

2.13 g/cm3 at 20 °C

anion traces

carbonate (as Na2CO3): ≤1%
chloride (Cl-): ≤0.012%
phosphate (PO43-): ≤0.0005%
silicate (as SiO2): ≤0.001%
sulfate (SO42-): ≤0.010%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ca: ≤0.0005%
Cu: ≤0.0002%
Fe: ≤0.0005%
K: ≤0.05%
Mg: ≤0.0005%
Ni: ≤0.00025%
Pb: ≤0.0005%
Zn: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium hydroxide (caustic soda) is an inorganic source of hydroxide ions that can be used to alkylate phenols, alcohols, aldehydes, and ketones. NaOH can also be used to convert amides into amines, to produce dichlorocarbenes, and in saponification of carboxylic acid derivatives.

Application

Sodium hydroxide may be used:
  • As a deprotonator in the synthesis of nanoparticles of zeolitic imidazolate framework (ZIF-94).
  • As a precipitating agent in the synthesis of NiO (nickel oxide) nanoparticles using nickel nitrate.
  • As a pH controller in the synthesis of zinc chromite (ZnCr2O4) oxide powder from zinc nitrate and chromium nitrate by hydrothermal process.

Analysis Note

Assay (acidimetric,NaOH): ≥ 99.0 %
Carbonate (as Na₂CO₃): ≤ 1.0 %
Chloride (Cl): ≤ 0.012 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.010 %
Total nitrogen (N): ≤ 0.0003 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.05
Mg (Magnesium): ≤ 0.0005 %
Ni (Nickel): ≤ 0.00025 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination of ammonium ions in drinking and surface water samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.

Photometric determination of ammonium ions in drinking and surface water samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.

Photometric determination of ammonium ions in drinking and surface water samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.

Photometric determination of ammonium ions in drinking and surface water samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service