Skip to Content
Merck
All Photos(2)

Documents

850795C

Avanti

18:3 PE

1,2-dilinolenoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1,2-di-(9Z,12Z,15Z-octadecatrienoyl)-sn-glycero-3-phosphoethanolamine; PE(18:3(9Z,12Z,15Z)/18:3(9Z,12Z,15Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H70NO8P
CAS Number:
Molecular Weight:
735.97
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850795C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850795C

concentration

10 mg/mL (850795C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\C/C=C\CC)=O)COC(CCCCCCC/C=C\C/C=C\C/C=C\CC)=O

General description

Phosphatidylethanolamine (PE) is a membrane phospholipid. The types of PE depends on the combination of free fatty acids on the α and β position. PE represents the main phospholipid class bacteria and the second most abundant in eukaryotic cells. PE is zwitterionic. PE is synthesized by Kennedy pathway.

Application

18:3 PE has been used in the preparation of liposomes. It may be suitable for use in the characterization of contact angle in monolayer formation.

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a key role in autophagy, apoptosis, membrane fusion, cell cycle and cognitive function. PE is essential for maintaining membrane architecture. PE functions to monitor proper progression of cytokinesis during cell division. PE is the precursor for many biologically active signaling molecules like diacylglycerol and phosphatidic acid.

Packaging

5 mL Clear Glass Sealed Ampule (850795C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomimetic membrane platform: fabrication, characterization and applications
Yildiz AA, et al.
Colloids and Interface Science Communications, 103, 510-516 (2013)
Ahu Arslan Yildiz et al.
Colloids and surfaces. B, Biointerfaces, 103, 510-516 (2012-12-25)
A facile method for assembly of biomimetic membranes serving as a platform for expression and insertion of membrane proteins is described. The membrane architecture was constructed in three steps: (i) assembly/printing of α-laminin peptide (P19) spacer on gold to separate
Force-from-lipids gating of mechanosensitive channels modulated by PUFAs
Ridone P, et al.
Journal of the Mechanical Behavior of Biomedical Materials, 79, 158-167 (2018)
Diarachidonoylphosphoethanolamine induces necrosis/necroptosis of malignant pleural mesothelioma cells
Kaku Y, et al.
Cellular Signalling, 27(9), 1713-1719 (2015)
The Kennedy pathway?de novo synthesis of phosphatidylethanolamine and phosphatidylcholine
Gibellini F and Smith TK
IUBMB Life, 62(6), 414-428 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service