Skip to Content
Merck
All Photos(1)

Key Documents

850483C

Avanti

18:1 Cy5 PC

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphocholine-N-(Cyanine 5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H124ClN4O9P
CAS Number:
Molecular Weight:
1316.26
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

99% (TLC)

form

solution

packaging

package of 1 × 250 μg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL in chloroform

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(CCNC(CCCCC[N+]1=C(/C=C/C=C/C=C2C(C)(C)C(C=CC=C3)=C3N\2C)C(C)(C)C4=C1C=CC=C4)=O)C)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Cl-]

InChI

1S/C77H123N4O9P.ClH/c1-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43-57-74(83)87-64-66(90-75(84)58-44-37-35-33-31-29-27-25-23-21-19-17-15-13-11-2)65-89-91(85,86)88-63-62-81(8,9)61-59-78-73(82)56-42-39-49-60-80-70-53-48-46-51-68(70)77(5,6)72(80)55-41-38-40-

InChI key

FRDSIEVJUUFAKX-IEXYBMNJSA-N

Packaging

5 mL Clear Glass Sealed Ampule (850483C-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Naomi Benne et al.
Advanced healthcare materials, 9(10), e2000043-e2000043 (2020-04-25)
Atherosclerosis is characterized by the retention of lipids in foam cells in the arterial intima. The liver X receptor (LXR) agonist GW3965 is a promising therapeutic compound, since it induces reverse cholesterol transport in foam cells. However, hepatic LXR activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service