Skip to Content
Merck
All Photos(3)

Documents

840055C

Avanti

Liver PC

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylcholine (Liver, Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840055C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840055C-25mg)

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid found in mammalian membranes. It consists of a positively charged quaternary amine in the headgroup and a negatively charged phosphate group.

Application

Liver PC is suitable for use:
  • in the preparation of two dimensional crystals of microsomal glutathione transferase 1 for electron microscopy
  • in the preparation of ′golgi lipid′ mixture for liposome recruitment assay
  • in lipid vesicle preparation for ligand transfer assay

Biochem/physiol Actions

Phosphatidylcholine (PC) is an important source of diacylglycerol and other signaling molecules. Liver PC has a saturated to unsaturated fatty acid ratio of 0.91.

Packaging

5 mL Clear Glass Sealed Ampule (840055C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
Stress sensor triggers conformational response of the integral membrane protein microsomal glutathione transferase 1
Busenlehner LS, et al.
Biochemistry, 43(35), 11145-11152 (2004)
GORAB scaffolds COPI at the trans-Golgi for efficient enzyme recycling and correct protein glycosylation
Witkos TM, et al.
Nature Communications, 10(1), 127-127 (2019)
The contribution of surface residues to membrane binding and ligand transfer by the alpha-tocopherol transfer protein (alpha-TTP)
Zhang WX, et al.
Journal of molecular biology, 405(4), 972-988 (2011)
Effect of bilayer phospholipid composition and curvature on ligand transfer by the alpha-tocopherol transfer protein
Zhang WX, et al.
Lipids, 44(7), 631-631 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service