Skip to Content
Merck
All Photos(2)

Key Documents

810198C

Avanti

18:1 NBD PS

Avanti Research - A Croda Brand 810198C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phospho-L-serine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H85N6O13P
CAS Number:
Molecular Weight:
985.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810198C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810198C

concentration

1 mg/mL (810198C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1 NBD PS is a fluorescent analog of phosphotidylserine (PS). Phosphotidylserine is considered as a vital lipid and is found in the inner mitochondrial region.

Application

NBD PS might be suitable as a component in premixed lipid solution for V-SNARE (N-ethylmaleimide-sensitive factor attachment protein receptor) proteo-liposomes preparation. It is also suitable for the preparation of control PS/PC (1-palmitoyl-2-oleoylsn-glycero-3-phospho-L-serine/1-palmitoyl-2-oleoyl-sn-glycero-phosphocholine) liposomes.

Biochem/physiol Actions

18:1 NBD PS is useful in labelling mitochondria. Phosphotidylserine is associated with blood clotting mechanism.

Packaging

5 mL Amber Glass Screw Cap Vial (810198C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PS (PhosphatidylSerine) Nature's Brain Booster A Vital Lipid Nutrient For Memory Mood and Stress, 84-84 (2005)
Nadège Bellance et al.
International journal of molecular sciences, 21(4) (2020-02-23)
Doxorubicin (DXR) is a drug widely used in chemotherapy. Its mode of action is based on its intercalation properties, involving the inhibition of topoisomerase II. However, few studies have reported the mitochondrial effects of DXR while investigating cardiac toxicity induced
Soluble N-Ethylmaleimide-Sensitive Factor Attachment Protein Receptor-Derived Peptides for Regulation of Mast Cell Degranulation.
Yang Y, et al.
Frontiers in Immunology (2018)
Soluble N-Ethylmaleimide-Sensitive Factor Attachment Protein Receptor-Derived Peptides for Regulation of Mast Cell Degranulation.
Yang Y, et al.
Frontiers in immunology (2018)
Mitochondria Supply Membranes for Autophagosome Biogenesis during Starvation
Hailey DW, et al.
Cell, 141(4), 656-667 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service