Skip to Content
Merck
All Photos(2)

Key Documents

700070P

Avanti

trihydroxycholestanoic acid

Avanti Research - A Croda Brand

Synonym(s):

3α,7α,12α-trihydroxycholestanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O5
CAS Number:
Molecular Weight:
450.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700070P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bile acids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]2[C@@H]([C@@]4([C@H](C1)C[C@@H](CC4)O)C)C[C@@H]([C@]3([C@H]2CC[C@@H]3[C@@H](CCCC(C)C(=O)O)C)C)O

InChI

1S/C27H46O5/c1-15(6-5-7-16(2)25(31)32)19-8-9-20-24-21(14-23(30)27(19,20)4)26(3)11-10-18(28)12-17(26)13-22(24)29/h15-24,28-30H,5-14H2,1-4H3,(H,31,32)/t15-,16?,17+,18-,19-,20+,21+,22-,23+,24+,26+,27-/m1/s1

InChI key

CNWPIIOQKZNXBB-VCVMUKOKSA-N

General description

Trihydroxycholestanoic acid (THCA) is an oxidative metabolite of cholesterol and a precursor to bile acids. Very long-chain acyl-CoA synthetase (VLCS) associated with endoplasmic reticulum and peroxisomes activates THCA, the C27 bile acid intermediate.[1]

Application

Trihydroxycholestanoic acid has been used as a sterol to test its reactivity with human mincle protein.[2]

Biochem/physiol Actions

Trihydroxycholestanoic acid (THCA) and dihydroxycholestanoic (DHCA) accumulation is observed in Zellweger syndrome and cholestatic liver disease.[3] The synthesis of (25R)-isomer of DHCA and THCA is favored by mitochondrial 27-hydroxylase.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700070P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ferdinandusse et al.
Journal of lipid research, 42(1), 137-141 (2001-02-13)
We identified a new peroxisomal disorder caused by a deficiency of the enzyme alpha-methylacyl-coenzyme A (CoA) racemase. Patients with this disorder show elevated plasma levels of pristanic acid and the bile acid intermediates di- and trihydroxycholestanoic acid (DHCA and THCA)
James O'Byrne et al.
The Journal of biological chemistry, 278(36), 34237-34244 (2003-06-18)
Bile acid-CoA:amino acid N-acyltransferase (BACAT) catalyzes the conjugation of bile acids to glycine and taurine for excretion into bile. By use of site-directed mutagenesis and sequence comparisons, we have identified Cys-235, Asp-328, and His-362 as constituting a catalytic triad in
Ryoko Kiyotake et al.
The Journal of biological chemistry, 290(42), 25322-25332 (2015-08-25)
C-type lectin receptors (CLRs) are an emerging family of pattern recognition receptors that recognizes pathogens or damaged tissue to trigger innate immune responses. However, endogenous ligands for CLRs are not fully understood. In this study, we sought to identify an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service