Skip to Content
Merck
All Photos(1)

Key Documents

330710X

Avanti

Mouse SPLASH LIPIDOMIX Mass Spec Standard

Avanti Research - A Croda Brand

Synonym(s):

SPLASH LIPIDOMIX Mass Spec Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

form

solution (1:1 dichloromethane:methanol)

packaging

pkg of 1 × 1 mL (330710X-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

mass spectrometry (MS): suitable

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Components

2 mL amber glass sealed ampule
The Mouse SPLASH LIPIDOMIX Mass Spec Standard is supplied in 1 mL of (1:1) dichloromethane:methanol and contains the following components:

Mixture ComponentTarget Conc. (μM)
  • 15:0-18:1 (d7) PC100
  • 15:0-18:1 (d7) PE7
  • 15:0-18:1 (d7) PS (Na salt)20
  • 15:0-18:1 (d7) PG (Na salt)5
  • 15:0-18:1 (d7) PI (NH4 salt)20
  • 15:0-18:1 (d7) PA (Na salt)10
  • 18:1-(d7) Lyso PC45
  • 18:1-(d7) Lyso PE2
  • 18:1-(d7) cholesterol ester250
  • C18(plasm)-18:1(d9) PC20
  • 15:0-18:1-(d7) DAG15
  • 15:0-18:1-(d7):15:0 TAG35
  • d18:1-18:1-(d9) SM20
  • C18(plasm)-18:1(d9) PE5

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 791637C15:0-18:1-d7-PC, Avanti Research - A Croda Brand 100μMSDS

  • 791638C15:0-18:1-d7-PE, Avanti Research - A Croda Brand 7μMSDS

  • 791639C15:0-18:1-d7-PS, Avanti Research - A Croda Brand 20μMSDS

  • 791640C15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C 5μMSDS

  • 791641C15:0-18:1-d7-PI, Avanti Research - A Croda Brand 20μMSDS

  • 791642C15:0-18:1-d7-PA, Avanti Research - A Croda Brand 10μMSDS

  • 791643C18:1-d7 Lyso PC, Avanti Research - A Croda Brand 45μMSDS

  • 791644C18:1-d7 Lyso PE, Avanti Research - A Croda Brand 791644C 2μMSDS

  • 852475CC18(plasm)-18:1(d9) PC, Avanti Research - A Croda Brand, chloroform solution 20μMSDS

  • 860740L18:1 SM (d18:1/18:1)-d9, Avanti Research - A Croda Brand 860740L-1MG
    860746L-1MG 20μM
    SDS

  • 852474CC18(plasm)-18:1(d9) PE, Avanti Research - A Croda Brand, chloroform solution 5μMSDS

See All (11)

also commonly purchased with this product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Valentina Bosello Travain et al.
Free radical biology & medicine, 147, 80-89 (2019-12-21)
GPx8 is a glutathione peroxidase homolog inserted in the membranes of endoplasmic reticulum (ER), where it seemingly plays a role in controlling redox status by preventing the spill of H2O2. We addressed the impact of GPx8 silencing on the lipidome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service