Merck
All Photos(4)
Key Documents
Z556734
Sigma-Aldrich
Aldrich® Benchclamp 2-prong
dual adjustment, size M
Synonym(s):
bench clamps
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
Pricing and availability is not currently available.
size
M
L × diam.
127 mm × 11 mm , arm
jaw capacity
0-75 mm
prong W
23 mm
General description
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)