Key Documents
Z408476
Sigma-Aldrich
Scienceware® Vibrasorb® vibration damping mount
size 13 3/4 in. × 13 3/4 in.
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18386-0001
size
13 3/4 in. × 13 3/4 in.
weight
42 lb
General description
Isolates equipment from disturbing vibrations down to 9 Hz. Molded of dense, 2 ½ in. thick, black and white terrazzo with a polished surface that resists scratches and chemicals. Supported by four Vibro-Absorber cushions and neoprene feet. Overall height is 3 in. Supports 16 kg.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vibrasorb is a registered trademark of SP Scienceware, a division of SP