Key Documents
Z288977
Supelco
Hamilton® needles for LT and TLL syringes
Kel-F Hub (KF), needle size 22s ga, needle L 51 mm (2 in.), pkg of 6 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Pricing and availability is not currently available.
material
blunt tip stainless steel needle (point style 3)
needle size
22s ga
feature
needle blunt tip
Kel-F Hub (KF)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 90534
needle L
51 mm (2 in.)
compatibility
for use with Hamilton LT and TLL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)