Key Documents
Z283770
Optifix® PTFE thread adapter
neck thread size GL 33
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
Pricing and availability is not currently available.
neck thread size
GL 33
manufacturer/tradename
Fortuna Graf
General description
Made of PTFE. Adapts Optifix dispensers to other bottle thread sizes.
Other Notes
Fits Aldrich pour-out bottles
Legal Information
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Is part #Z283770 Optifix PTFE thread adapter compatible with strong acids
    1 answer
    1. Polytetrafluoroethylene (PTFE) is highly resistant to most acids and is listed as having 'excellent' compatibility with commonly used concentrated acids, such as Acetic, Hydrochloric, and Nitric acids. Please see the link below to review additional chemical compatibility data: https://www.calpaclab.com/teflon-ptfe-compatibility/?srsltid=AfmBOoqz2vz-h1mq5PeFKhcgAP3LCoCiLm_TT8d392M0LTwvd7N079Ym
      Helpful?