The maximum inlet pressure is 3,000 psig.
Z146706
Aldrich® non-corrosive lecture-bottle gas regulator
size, CGA Inlet size 180
Synonym(s):
Gas control valve
About This Item
Recommended Products
measuring range
0-90 psig
CGA Inlet size
180
size
General description
- Single-stage
- Inlet: CGA 180
- Outlet: 1/4" male NPTF
- Max. inlet pressure: 3,000 psig
- Temp. range: -40 to 140°F
- Compact chrome-plated brass body
- Lecture-bottle gas regulators include a PTFE sealing washer
- Order spare PTFE sealing washers separately
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
-
Could you tell me the maximum ingoing pressure
1 answer-
Helpful?
-
-
Does lecture bottle regulator Z146706 have a CRN number?
1 answer-
No, this lecture bottle regulator does not contain a CRN number.
Helpful?
-
-
What are the thread dimensions of a lecture-bottle adapter?
1 answer-
Product Z146706 is a single-stage lecture bottle regulator. It is not an adapter. The inlet is a CGA 180 and the outlet is a 1/4" male NPTF. See the image below for the product drawing.
As adapters vary in size, additional information would be required in order to answer your question. To review the available lecture bottle adapters see the link below:
https://www.sigmaaldrich.com/search/lecture-bottle-adapter?focus=products&page=1&perpage=30&sort=relevance&term=lecture%20bottle%20adapter&type=productTo discuss further, please navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquiries" under the Products Section with all the required information, including the product number of interest, so that a member of the Technical Service team can reach out to assist further. Thank you.
Helpful?
-
-
What is the maximum delivery pressure for your Z146706 gas regulator?
1 answer-
Maximum inlet pressure is 3,000 psig.
Helpful?
-