Key Documents
TEM-CF400CU
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
size 400 mesh, box of 25
Synonym(s):
CARBON FILM 400# COPPER 25/PK
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
400 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Copper grid, 400 mesh.