Key Documents
TEM-CF200CU
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
size 200 mesh, box of 25
Synonym(s):
CARBON FILM 200 MESH CU 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Copper grid, 200 mesh.