Key Documents
SYND151814
Synthware short path distillation head
joint: ST/NS 14/20
Synonym(s):
Synthware D151814, Short path head
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D151814
Related Categories
General description
Short-path design for microscale application, used with 25 mm immersion thermometer, and D27 distillation receiver.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)